Phenol red sodium salt ACS

Supplier: Thermo Scientific Chemicals
Warning

41725-0050 41725-0100 41725-1000
TS41725-0050EA 33 USD
TS41725-0050 TS41725-0100 TS41725-1000
Phenol red sodium salt ACS
Phenol red sodium salt
Formula: C₁₉H₁₃NaO₅S
Melting Pt: 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS Number: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397

Order Now

Specification Test Results

Appearance Dark red to red-brown Powder
Infrared spectrum Conforms
Visual transition interval From pH 6.8 (yellow) to pH 8.2 (red)
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR