New methylene Blue N, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Basic Blue 24Methylene Blue N

41553-0025
TS41553-0025EA 88.3 USD
TS41553-0025
New methylene Blue N, pure
New methylene Blue N
Formula: C₁₈H₂₂ClN₃S
MW: 347.91 g/mol
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00135878
CAS Number: 1934-16-3

Order Now

Specification Test Results

Appearance Green to dark brown Crystalline powder or crystals
Infrared spectrum Conforms
Dye content ca 70%
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR