Morpholine 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

41516-5000 41516-0050 41516-0025
TS41516-0050EA 22.36 USD
TS41516-0050 TS41516-5000 TS41516-0025
Morpholine 99+% ACS
Morpholine
Formula: C₄H₉NO
MW: 87.12 g/mol
Boiling Pt: 128.9 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.99 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00005972
CAS Number: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8,I
https://us.vwr.com/stibo/web/std.lang.all/82/95/2298295.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4530 to 1.4550 (20°C, 589 nm)
Boiling point 126.0 to 130.0°C
Color scale ≤15 APHA
Titration with HCl ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR