Methyl violet 2B, pure, certified

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride

41507-0250 41507-1000 41507-5000
TS41507-0250EA 33 USD
TS41507-0250 TS41507-1000 TS41507-5000
Methyl violet 2B, pure, certified
Methyl Violet 2B
Formula: C₂₄H₂₈ClN₃
MW: 393.96 g/mol
Melting Pt: 137
Storage Temperature: Ambient
MDL Number: MFCD00012568
CAS Number: 8004-87-3
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance Green Crystalline powder
Infrared spectrum Conforms
Dye content ≥75 %
UV Lambda max 582 to 588 nm
Absorption Ratio P-15/P+15 1.06 to 1.19
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR