Hematoxylin

Supplier: Thermo Scientific Chemicals

Synonyms: (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaolHydroxybrasilinNSC 2785DcH-48Hydroxybrazilin(+)-HematoxylinNatural Black I

41118-0250
TS41118-0250EA 67 USD
TS41118-0250
Hematoxylin
Hematoxylin
Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
Melting Pt: 291…293 °C
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2
EINECS: 208-237-3
Merck Index: 13,04654

Order Now

Specification Test Results

Appearance Beige to orange to brown Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (1 g, 105 °C, 3 hrs)
UV spectrophotometry ≥80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR