Erythrosin B disodium salt, pure, certified

Supplier: Thermo Scientific Chemicals
Warning

40945-0250 40945-1000
TS40945-0250EA 61 USD
TS40945-0250 TS40945-1000
Erythrosin B disodium salt, pure, certified
Erythrosin B disodium salt
Formula: C₂₀H₆I₄Na₂O₅
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00144257
CAS Number: 16423-68-0
EINECS: 240-474-8

Order Now

Specification Test Results

Appearance Red to brown Powder
Infrared spectrum Conforms
Dye content ≥80 %
UV Lambda max 524 to 527 nm
Absorption Ratio P-15/P+15 1.15 to 1.55
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR