Phosphorus pentoxide - methanesulfonic acid

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Eaton's Reagent

36980-0025 36980-5000
TS36980-5000EA 164 USD
TS36980-5000 TS36980-0025
Phosphorus pentoxide - methanesulfonic acid
Phosphorus pentoxide - methanesulfonic acid
MW: 141.95 g/mol
Boiling Pt: 122 °C (1 mmHg)
Density: 1.5 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00070545
CAS Number: 39394-84-8
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance to light brown or pale pink Solution (May contain precipitate, re-dissolves on warming)
Refractive index 1.4325 to 1.4375 (20°C, 589 nm)
Titration with NaOH 6.5 to 10.0 % (P2O5)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR