1-Butanethiol 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
22062-0010
22062-0050
22062-0500
22062-2500
TS22062-0010EA
113
USD
TS22062-0010
TS22062-0050
TS22062-0500
TS22062-2500
1-Butanethiol 99+%
1-Butanethiol
Formula:
C₄H₁₀S Boiling Pt: 98 °C (760 mmHg) Melting Pt: –116 °C Density: 0.84 g/cm³ Flash Pt: 0.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004905 CAS Number: 109-79-5 EINECS: 203-705-3 UN: 2347 ADR: 3,II Merck Index: 12,01611 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4420 to 1.4440 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...