Benzyltrimethylammonium chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium chloride

21186-0010 21186-0500 21186-2500
TS21186-0010EA 162 USD
TS21186-0010 TS21186-0500 TS21186-2500
Benzyltrimethylammonium chloride 98+%
Benzyltrimethylammonium chloride
Formula: C₁₀H₁₆ClN
Melting Pt: 239 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011782
CAS Number: 56-93-9
EINECS: 200-300-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to cream Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR