4-Nitrophenol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Hydroxy-4-nitrobenzene, p-Nitrophenol

15705-0010 15705-0050 15705-0051 15705-1000 15705-2500
TS15705-0010EA 101 USD
TS15705-0010 TS15705-0050 TS15705-0051 TS15705-1000 TS15705-2500
4-Nitrophenol 99%
4-Nitrophenol
Formula: C₆H₅NO₃
Boiling Pt: 279 °C (760 mmHg)
Melting Pt: >112 °C
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00007331
CAS Number: 100-02-7
EINECS: 202-811-7
UN: 1663
ADR: 6.1,III
Merck Index: 13,06654

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 112 to 115 °C
GC ≥98.5 %
Titration with NaOH ≥96.0 to ≤104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR