Light green SF yellowish, pure, high purity biological stain

Supplier: Thermo Scientific Chemicals

22977-0100 22977-1000
TS22977-0100EA 65 USD
TS22977-0100 TS22977-1000
Light green SF yellowish, pure, high purity biological stain
Light Green SF Yellowish
Formula: C₃₇H₃₄N₂Na₂O₉S₃
Melting Pt: 288 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012121
CAS Number: 5141-20-8
EINECS: 225-906-5
Merck Index: 1305506

Order Now

Specification Test Results

Appearance Purple Powder or crystals
Infrared spectrum Conforms
UV spectrum Complies
Loss on drying ≤10 % (1 g, 110°C, 2 h)
UV Lambda max 629 to 634 nm (H2O, 5 mg/l, 1 cm)
Absorption Ratio P-15/P+15 0.96 to 1.16
Titration ≥60 % (As is)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR