Brilliant green, pure, high purity biological stain

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulfate, Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Brilliant Green hydrogen sulfate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate

22960-1000 22960-5000
TS22960-1000EA 74 USD
TS22960-1000 TS22960-5000
Brilliant green, pure, high purity biological stain
Brilliant Green
Formula: C₂₇H₃₄N₂O₄S
MW: 482.65 g/mol
Melting Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00011880
CAS Number: 633-03-4
EINECS: 211-190-1

Order Now

Specification Test Results

Appearance Green to green-yellow Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤5 % (105°C)
UV Lambda max 625 to 631 nm (in 50% ethanol)
Absorption Ratio P-15/P+15 0.94 to 1.14
Specific absorbance A (1%/1cm) ≥1900 (at 625 to 631 nm)
Visual transition interval from pH 0.0 (yellow) to pH 2.6 (green)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR