EDTA dipotassium salt dihydrate 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethylenediaminetetraacetic acid dipotassium salt dihydrate
, Ethylenedinitrilotetra-acetic acid dipotassium salt dihydrate
, Ethylenedinitrilotetraacetic acid dipotassium salt
, Titriplex® Potassium salt
, Edetic acid dipotassium salt dihydrate
, EDTA dipotassium dihydrate
, Dipotassium dihydrogen ethylenediaminetetraacetate dihydrate
, Potassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate
, Ethylenediaminetetra-acetic acid dipotassium salt dihydrate
20974-0025
20974-1000
20974-5000
TS20974-1000EA
72
USD
TS20974-1000
TS20974-0025
TS20974-5000
EDTA dipotassium salt dihydrate 99%, Extra Pure
EDTA dipotassium salt dihydrate
Formula:
C₁₀H₁₄K₂N₂O₈·2H₂O MW: 404.46 g/mol Melting Pt: 272 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150036 CAS Number: 25102-12-9 EINECS: 217-895-0 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration Complexometric | ≥98.5 % |
Solubility | (10 % in water) Clear colorless |
pH | 4 to 5 (5 % in water) |
Chloride (Cl) | ≤50 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...