Evans blue

Supplier: Thermo Scientific Chemicals
Danger

19555-0050 19555-0250
TS19555-0050EA 40 USD
TS19555-0050 TS19555-0250
Evans blue
Evans blue
Formula: C₃₄H₂₈N₆O₁₄S₄
MW: 960.79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004021
CAS Number: 314-13-6

Order Now

Specification Test Results

Appearance Dark brown Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤10 %
UV Lambda max 605 to 611 nm
Specific extinction E (1%/1cm) ≥710 (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR