Basic Blue 24 (New Methylene Blue), pure

Supplier: Thermo Scientific Chemicals

Synonyms: New Methylene Blue NNew Methylene Blue N, zinc chloride double salt

19202-0100
TS19202-0100EA 104 USD
TS19202-0100
Basic Blue 24 (New Methylene Blue), pure
Basic Blue 24 (New Methylene Blue)
Formula: C₁₈H₂₂ClN₃S·0.5 ZnCl₂
MW: 416.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078135
CAS Number: 6586-05-6

Order Now

Specification Test Results

Appearance Dark brown to deep purple Crystalline powder
Infrared spectrum Conforms
Dye content ≥90 %
UV Lambda max 628 to 634 nm (c=5 mg/l, water)
UV Lambda max 588 to 594 nm (c=5 mg/l, water)
UV Lambda max 284 to 290 nm (c=5 mg/l, water)
UV Lambda max 242 to 248 nm (c=5 mg/l, water)
Molar extinction coefficient ≥40000 (c=5 mg/l, water) 628 to 634 nm
Molar extinction coefficient ≥36000 (c=5 mg/l, water) 588 to 594 nm
Molar extinction coefficient ≥28000 (c=5 mg/l, water) 284 to 290 nm
Molar extinction coefficient ≥16000 (c=5 mg/l, water) 242 to 248 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR