Tropaeolin O, pure

Supplier: Thermo Scientific Chemicals

Synonyms: CI 14270, Sodium 2,4-Dihydroxyazobenzene-4'-sulfonate, Tropaeolin O sodium salt, Sulpho orange, Chrysoin, Sulfo orange, 2,4-Dihydroxyazobenzene-4'-sulphonic acid sodium salt, Sodium 2,4-Dihydroxyazobenzene-4'-sulphonate, 2,4-Dihydroxyazobenzene-4'-sulfonic acid sodium salt, Acid Orange 6, Resorcinol Yellow

19018-0250 19018-1000
TS19018-0250EA 75 USD
TS19018-0250 TS19018-1000
Tropaeolin O, pure
Tropaeolin O
Formula: C₁₂H₉N₂O₅SNa
MW: 316.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007499
CAS Number: 547-57-9

Order Now

Specification Test Results

Appearance Rust brown to dark brown Powder
Infrared spectrum Conforms
Solubility (in water) Passes test
Visual transition interval from pH 11.1 (yellow) to pH 12.7 (orange)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR