Sudan black B, high purity biological stain
Supplier: Thermo Scientific Chemicals
19016-0250
19016-1000
TS19016-0250EA
85
USD
TS19016-0250
TS19016-1000
Sudan black B, high purity biological stain
Sudan black B
Formula:
C₂₉H₂₄N₆ Melting Pt: 120…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006919 CAS Number: 4197-25-5 EINECS: 224-087-1 |
Specification Test Results
Appearance | Dark brown to black Crystalline powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...