Trypan blue
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt
, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt
, CI 23850
, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulfonate)
, Direct Blue 14
, AzidinBlue
, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
, Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulfonic acid tetrasodium salt
, Benzamine blue
, Trypan blue sodium salt
, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulfonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt
, Diphenyl Blue
Formula:
C₃₄H₂₄N₆Na₄O₁₄S₄ MW: 960.82 g/mol Melting Pt: >280 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003969 CAS Number: 72-57-1 EINECS: 200-786-7 |
Specification Test Results
Appearance | Dark blue to brown-black Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Dye content | ≥60 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...