Starch, soluble for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: StarchVitex® iodine indicatorAmylodextrin

17713-2500 17713-0050 17713-0010
TS17713-2500EA 46.52 USD
TS17713-2500 TS17713-0010 TS17713-0050
Starch, soluble for analysis
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Melting Pt: 256…258 °C
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 1308878

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤10 %
Sulfated ash ≤0.5 %
Identification Positive
pH 5 to 7.5 (2 % at 25°C)
Sensitivity Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR