Trifluoromethanesulphonic anhydride 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Triflic anhydride

17506-0010 17506-0500 17506-2500
TS17506-0010EA 39 USD
TS17506-0010 TS17506-0500 TS17506-2500
Trifluoromethanesulphonic anhydride 98+%
Trifluoromethanesulphonic anhydride
Formula: C₂F₆O₅S₂
Boiling Pt: 80 °C (760 mmHg)
Density: 1.7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000408
CAS Number: 358-23-6
EINECS: 206-616-8
UN: 3265
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3200 to 1.3230 (20°C, 589 nm)
Titration with CHA 98.0 to 101.0 %
Impurity ≤1.0 % CF3SO3H
Distillation range ≥95 % (between 123-133°C)
Pour point < -40°C
Residue ≤10 ug/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR