2,4,6-Tris(2-pyridyl)-1,3,5-triazine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4,6-Tri(2-pyridyl)-s-triazine, 2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 2,4,6-Tris(2-pyridyl)-s-triazine

16807-0010 16807-0050 16807-0250 16807-1000
TS16807-0010EA 40 USD
TS16807-0010 TS16807-0050 TS16807-0250 TS16807-1000
2,4,6-Tris(2-pyridyl)-1,3,5-triazine 99%
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
Formula: C₁₈H₁₂N₆
MW: 312.33 g/mol
Melting Pt: 248…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00006045
CAS Number: 3682-35-7
EINECS: 222-965-9

Order Now

Specification Test Results

Appearance White to cream to yellow Powder
Infrared spectrum Conforms
Melting point 248 to 252 °C
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR