Bathophenanthroline 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)

15832-0010 15832-0050 15832-0250
TS15832-0010EA 115 USD
TS15832-0010 TS15832-0050 TS15832-0250
Bathophenanthroline 99%
Bathophenanthroline
Formula: C₂₄H₁₆N₂
MW: 332.4 g/mol
Boiling Pt: 508
Melting Pt: 218…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00004976
CAS Number: 1662-01-7
EINECS: 216-767-1

Order Now

Specification Test Results

Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
Melting point 218 to 222 °C
Titration with HClO4 ≥98.5 %
Suitability (for Fe determination) Passes test
UV at 280 nm A: ≤0.01
Color scale ≤15 APHA (0.1 M in water)
pH 5.0 to 9.0
Heavy metals (as Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR