Methyl orange, pure indicator

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium saltMethyl orange sodium saltHelianthine4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulfonic acid, sodium salt

15142-1000 15142-5000
TS15142-5000EA 128 USD
TS15142-5000 TS15142-1000
Methyl orange, pure indicator
Methyl orange
Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Boiling Pt: ∼90 °C (1013 hPa)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1,III
Merck Index: 1306127

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Solubility (0.1 % in water) Clear
UV Lambda max 461 to 469 nm (in 0.01N NaOH)
Visual transition interval from pH 3.2 (pink or red)to pH 4.4 (yellow-orange)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR