Methyl red sodium salt, pure indicator

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate

15141-0250 15141-1000 15141-5000
TS15141-0250EA 41 USD
TS15141-0250 TS15141-1000 TS15141-5000
Methyl red sodium salt, pure indicator
Methyl red sodium salt
Formula: C₁₅H₁₄N₃NaO₂
Storage Temperature: Ambient
MDL Number: MFCD00002426
CAS Number: 845-10-3
EINECS: 212-682-9
Merck Index: 1306142

Order Now

Specification Test Results

Appearance Red to brown Powder
Infrared spectrum Conforms
Solubility (0.1% in water) clear to faint turbid solution
Visual transition interval from pH 4.2 (red) to pH 6.2 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR