Acetyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acetyl chloride 1M in Dichloromethane

15127-0010 15127-0025
TS15127-0010EA 112 USD
TS15127-0010 TS15127-0025
Acetyl chloride 98%
Acetyl chloride
Formula: C₂H₃ClO
MW: 78.5 g/mol
Boiling Pt: 50.9
Melting Pt: -112
Density: 1.1 g/cm³
Flash Pt: 4 °C
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 1717
ADR: 3 II
Merck Index: 13,00087

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3876 to 1.3896 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤30 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR