Trichloroacetonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

13937-0025 13937-0050 13937-1000 13937-5000
TS13937-0025EA 1342 USD
TS13937-0025 TS13937-0050 TS13937-1000 TS13937-5000
Trichloroacetonitrile 98%
Trichloroacetonitrile
Formula: C₂Cl₃N
MW: 144.39 g/mol
Boiling Pt: 85.7
Melting Pt: -44
Density: 1.44 g/cm³
Flash Pt: 195 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001842
CAS Number: 545-06-2
EINECS: 208-885-7
UN: 3276
ADR: 6.1 II
Merck Index: 12,09757

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR