Ethylenetetracarbonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetracyanoethylene

13805-0050 13805-0100 13805-0250
TS13805-0050EA 69 USD
TS13805-0050 TS13805-0100 TS13805-0250
Ethylenetetracarbonitrile 98%
Ethylenetetracarbonitrile
Formula: C₆N₄
Boiling Pt: 223 °C (760 mmHg)
Melting Pt: 196…201 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001850
CAS Number: 670-54-2
EINECS: 211-578-0
UN: 3439
ADR: 6.1,I
Merck Index: 1209336

Order Now

Specification Test Results

Appearance White to beige to light brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 196 to 201 °C
Assay ≥97.5 % (Ex N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR