Piperazine 99%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
13129-0025
13129-0100
13129-1000
13129-5000
TS13129-0025EA
223
USD
TS13129-0025
TS13129-0100
TS13129-1000
TS13129-5000
Piperazine 99%, Extra Pure
Piperazine
Formula:
C₄H₁₀N₂ MW: 86.14 g/mol Boiling Pt: 146…148 °C (760 mmHg) Melting Pt: 107…111 °C Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005953 CAS Number: 110-85-0 EINECS: 203-808-3 UN: 2579 ADR: 8,III Merck Index: 1307545 |
Specification Test Results
Appearance | White to light yellow Crystalline flakes |
Infrared spectrum | Conforms |
Water | ≤1 % |
GC | ≥98.5 % |
Color of solution | ≤30 APHA (50% in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...