2,3-Dichloro-5,6-dicyano-p-benzoquinone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
DDQ, 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 2,3-Dichloro-5,6-dicyanoquinone, 4,5-Dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
11330-0100
11330-1000
11330-5000
TS11330-0100EA
51
USD
TS11330-0100
TS11330-1000
TS11330-5000
2,3-Dichloro-5,6-dicyano-p-benzoquinone 98%
2,3-Dichloro-5,6-dicyano-p-benzoquinone
Formula:
C₈Cl₂N₂O₂ Melting Pt: 210…217 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001593 CAS Number: 84-58-2 EINECS: 201-542-2 UN: 3439 ADR: 6.1,III Merck Index: 1303088 |
Specification Test Results
Appearance | Yellow to orange Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 210 to 217 °C |
Titration Iodometric | ≥97.5 % |
Loss on drying | ≤0.5 % (1 g, 80 °C, 2 h, 40 mmHg) (vacuum) |
Sulfated ash | ≤0.2 % (600 °C) |
Specific absorbance A (1%/1cm) | ≥1800 (at lambda max.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...