Bromotrichloromethane 99%

Supplier: Thermo Scientific Chemicals
Warning

10749-1000 10749-2500
TS10749-1000EA 79 USD
TS10749-1000 TS10749-2500
Bromotrichloromethane 99%
Bromotrichloromethane
Formula: BrCl₃C
Boiling Pt: 105 °C (1013 hPa)
Melting Pt: –6 °C
Density: 1.997 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000783
CAS Number: 75-62-7
EINECS: 200-886-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5055 to 1.5075 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR