Biuret 97%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Allophanic acid amide
, Carbamylurea
10652-0050
10652-0250
10652-1000
TS10652-0050EA
20.9
USD
TS10652-0050
TS10652-0250
TS10652-1000
Biuret 97%, Extra Pure
Biuret
Formula:
C₂H₅N₃O₂ MW: 103.08 g/mol Melting Pt: 187…194 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00007946 CAS Number: 108-19-0 EINECS: 203-559-0 Merck Index: 1301305 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 184 to 190 °C |
Assay | ≥96.0 % (ex N) |
Loss on drying | ≤1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...