Gentamicin sulfate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: NMD Inhibitor IIGaramycinGentiomycin CGentamicin sulfate saltGentamicin sulphate salt

61398-0010
TS61398-0010EA 176.17 USD
TS61398-0010
Gentamicin sulfate
Gentamicin sulphate
Melting Pt: 218…237 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270181
CAS Number: 1405-41-0

Order Now

Specification Test Results

Appearance White Powder
Identification passes test
Specific optical rotation +107° to +121° (20 °C, 589 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR