Aluminium oxide basic Brockmann Grade I 40-300 µm, 60A for chromatography

Supplier: Thermo Scientific Chemicals

18999-0010 18999-0051 18999-0250
TS18999-0010EA 118 USD
TS18999-0010 TS18999-0051 TS18999-0250
Aluminium oxide basic Brockmann Grade I 40-300 µm, 60A for chromatography
Aluminium oxide basic
Formula: Al₂O₃
Boiling Pt: 2977 °C (760 mmHg)
Melting Pt: 2030 °C
Density: 3.97 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Particle size ≤5 % (<40 µm)
Specific surface area 185 to 225 m²/g
pH 8.5 to 10.5 (10% aqueous suspension)
Sodium oxide (Na2O) ≤0.4 %
Silicon dioxide (SiO2) ≤0.03 %
Iron(III)oxide (Fe2O3) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR