Petroleum spirit, 80…110 °C for analysis

Supplier: Thermo Scientific Chemicals
Danger

27823-0025 278230010 278230100
TS27823-0025EA 152 USD
TS27823-0025 TS278230010 TS278230100
Petroleum spirit, 80…110 °C for analysis
Petroleum spirit
Boiling Pt: 80…110 °C (760 mmHg)
Melting Pt: –40 °C
Density: 0.725 g/cm³
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3980 to 1.4060 (20°C, 589 nm)
Boiling point 78 to 113°C
Density (15°C) 0.719 to 0.729 kg/l
Color scale ≤10 APHA
Cadmium (Cd) ≤0.1 ppm
Cobalt (Co) ≤0.1 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.1 ppm
Nickel (Ni) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
n-Hexane ≤5 %
Benzene ≤20 ppm
Aromatics ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR