Celite® 545, filter aid
Supplier: Thermo Scientific Chemicals
Synonyms:
DE, Diatomaceous earth, flux-calcined
34967-0025
34967-0100
34967-5000
TS34967-0025EA
128
USD
TS34967-0025
TS34967-0100
TS34967-5000
Celite® 545, filter aid
Celite® 545, filter aid
Formula:
SiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00132803 CAS Number: 68855-54-9 EINECS: 272-489-0 |
Specification Test Results
Appearance | White to almost white Powder |
Particle size | ≤25 % (Retained on 150 mesh) |
Wet density | 255 to 370 g/l |
pH | 9.0 to 11.0 (10 % slurry) |
Permeability | 2.6 to 6.5 Darcy |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...