Silica gel 3-6 mm, non toxic for drying

Supplier: Thermo Scientific Chemicals

35740-0010 35740-0050 357400250
TS35740-0010EA 89 USD
TS35740-0010 TS35740-0050 TS357400250
Silica gel 3-6 mm, non toxic for drying
Silica gel
Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
REACH: 01-2119379499-16

Order Now

Specification Test Results

Appearance Colorless to white Irregular granules
Particle size 3.0 to 6.0 mm
Residual water ≤2.0 % (140°C, 4 h)
Water adsorption ≥20.0 % (40% rH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR