Polyethylene glycol 300

Supplier: Thermo Scientific Chemicals

Synonyms: PEG 300

19222-0010 192220100 19222-0025
TS19222-0010EA 95.87 USD
TS19222-0010 TS19222-0025 TS192220100
Polyethylene glycol 300
Polyethylene glycol 300
Formula: H(OCH₂CH₂)nOH
MW: 300 g/mol
Melting Pt: 15…10 °C
Density: 1.12 g/cm³
Flash Pt: >180 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4650 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
Color scale ≤50 APHA
Hydroxyl value 356 to 394 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR