Polyethyleneimine, M.W. ∼ 60,000 50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: PolyaziridinePolyethyleniminePolyethylenimine (branched)Polyaziridine Mw 25000PEICorcatPolymin PEthyleneimine PolymerAziridinePoly(ethyleneimine)

17857-0010 178572500 178571000
TS178571000EA 55.8 USD
TS178571000 TS178572500 TS17857-0010
Polyethyleneimine, M.W. ∼ 60,000 50% (w/w) in aqueous solution
Polyethyleneimine
Formula: (CH₂CH₂NH)n
Boiling Pt: 100 °C (760 mmHg)
Density: 1.07 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00084427
CAS Number: 9002-98-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Assay 48 to 52 % Solids
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR