Charcoal activated, NORIT® SA 2, decolorizing

Supplier: Thermo Scientific Chemicals

Synonyms: Activated carbonCarbon activatedActivated charcoal

404035000 40403-0025
TS40403-0025EA 202 USD
TS40403-0025 TS404035000
Charcoal activated, NORIT® SA 2, decolorizing
Charcoal activated
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652 °C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Specification Test Results

Appearance Black Powder
Loss on drying ≤10 %
Molasses number ≤390
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR