Charcoal activated, DARCO® 4-12 mesh, decolorizing

Supplier: Thermo Scientific Chemicals

Synonyms: Activated carbonCarbon activatedActivated charcoal

40399-0100 403990030 403995000
TS403990030EA 313 USD
TS403990030 TS40399-0100 TS403995000
Charcoal activated, DARCO® 4-12 mesh, decolorizing
Charcoal activated
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652 °C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Specification Test Results

Appearance Black Granules and/or chunks
Loss on drying ≤12 %
Particle size ≤5 % (> 4 mesh)
Particle size ≤5 % (< 12 mesh)
Particle size ≤0.4 % (dust)
pH 3.5 to 6

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR