Copper(II) sulfate, anhydrous 98%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
42287-0025
AC42287-0050
AC42287-0100
AC42287-1000
AC42287-5000
TS42287-0025EA
663
USD
TS42287-0025
TS42287-0050
TS42287-0100
TS42287-1000
TS42287-5000
Copper(II) sulfate, anhydrous 98%, pure
Copper (II) sulphate
Formula:
CuSO₄ Melting Pt: 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010981 CAS Number: 7758-98-7 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | Light gray Powder |
Titration Iodometric | ≥97.5 % |
Chloride (Cl) | ≤50 ppm |
Iron (Fe) | ≤100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...