L(-)-Glutathione (oxidised form) 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
L(-)-Glutathione, oxidized
, GSSG
, (2S,2'S)-5,5'-(((2R,2'R)-Disulphanediylbis(1-((carboxymethyl)amino)-1-oxopropane-3,2-diyl))bis(azanediyl))bis(2-amino-5-oxopentanoic acid)
, Bi(glutathion-S-yl)
, Glutathione oxidized
, Glutathiol
, Oxidized Glutathione (GSSG)
32022-0050
32022-5000
TS32022-5000EA
62.21
USD
TS32022-5000
TS32022-0050
L(-)-Glutathione (oxidised form) 98%
L(-)-Glutathione (oxidized form)
Formula:
C₂₀H₃₂N₆O₁₂S₂ MW: 612.64 g/mol Melting Pt: 178 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063106 CAS Number: 27025-41-8 EINECS: 248-170-7 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤8 % (105°C, 3 h) |
HPLC | ≥97.5 % |
Residue after ignition | ≤0.5 % |
Specific optical rotation | -92° to -106° (20°C, 589 nm) (c=4, water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...