n-Heptane 99+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

38379-0010 383790025 383790050 383790250
TS38379-0010EA 108 USD
TS38379-0010 TS383790025 TS383790050 TS383790250
n-Heptane 99+%, extra pure
n-Heptane
Formula: H₃C(CH₂)₅CH₃
Boiling Pt: 98 °C (760 mmHg)
Melting Pt: –91 °C
Density: 0.683 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥99.0 %
Residue after evaporation ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR