Heptane (mixture of isomers), Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
44427-0050
TS44427-0050EA
96.64
USD
TS44427-0050
Heptane (mixture of isomers), Technical Grade
Heptane (mixture of isomers)
Formula:
H₃C(CH₂)₅CH₃ MW: 100.2 g/mol Boiling Pt: 98 °C (760 mmHg) Melting Pt: –91 °C Density: 0.683 g/cm³ Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 UN: 1206 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Boiling point | 94 to 100°C |
Density | 0.685 to 0.730 kg/l (15°C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...