D-Glucoronic acid 98%

Supplier: Thermo Scientific Chemicals

30988-1000 30988-0100 30988-0250
TS30988-1000EA 483 USD
TS30988-1000 TS30988-0100 TS30988-0250
D-Glucoronic acid 98%
D-Glucoronic acid
Formula: C₆H₁₀O₇
Melting Pt: 158…162 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00166981
CAS Number: 6556-12-3
EINECS: 229-486-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Specific optical rotation +35.5° to +37.5° (20°C, 589 nm) (c=6, H2O) (after 4 h)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR