Sodium polyanetholesulfonate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Polyanetholesulfonic acid sodium salt, Polyanetholesulphonic acid sodium salt

41963-5000 41963-0250 41963-1000
TS41963-5000EA 1891 USD
TS41963-5000 TS41963-0250 TS41963-1000
Sodium polyanetholesulfonate
Sodium polyanetholesulfonate
Formula: (C₁₀H₁₁O₄SNa)n
Storage Temperature: Ambient
MDL Number: MFCD00148427
CAS Number: 55963-78-5

Order Now

Specification Test Results

Appearance White to yellow to beige to tan Powder or crystals
Infrared spectrum Conforms
pH 5.0 to 7.0 (1 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR