2-Aminoethyl methacrylate hydrochloride 90% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Aminoethyl 2-methylprop-2-enoate hydrochloride
35781-0250
35781-0050
TS35781-0250EA
342
USD
TS35781-0250
TS35781-0050
2-Aminoethyl methacrylate hydrochloride 90% stabilised
2-Aminoethyl methacrylate hydrochloride
Formula:
C₆H₁₂ClNO₂ MW: 165.617 g/mol Melting Pt: 116…118 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00078260 CAS Number: 2420-94-2 |
Specification Test Results
Appearance | White to light green to light brown Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥88 % |
Stabilizer | 500 to 1000 ppm phenothiazine |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...