Carbonylcyanide-3-chlorophenylhydrazone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: CCCP, Carbonyl cyanide m-chlorophenylhydrazone, Carbonyl cyanide m-chlorophenylhydrazone

22813-0010 22813-1000
TS22813-0010EA 178 USD
TS22813-0010 TS22813-1000
Carbonylcyanide-3-chlorophenylhydrazone 98%
Carbonylcyanide-3-chlorophenylhydrazone
Formula: C₉H₅ClN₄
MW: 204.617 g/mol
Melting Pt: 172…177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001848
CAS Number: 555-60-2
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Yellow to orange to orange-brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR