Triolein 99%

Supplier: Thermo Scientific Chemicals

36812-0050 36812-0010 36812-0250 36812-1000
TS36812-0050EA 93 USD
TS36812-0050 TS36812-0010 TS36812-0250 TS36812-1000
Triolein 99%
Triolein
Formula: C₅₇H₁₀₄O₆
Boiling Pt: 235…240 °C (18 mmHg)
Density: 0.913 g/cm³
Storage Temperature: Freezer
MDL Number: MFCD00137563
CAS Number: 122-32-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4680 to 1.4700 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR