Methyl-ɑ-D-mannopyranoside 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: ɑ-Methyl-D-mannoside, Methyl-ɑ-D-Mannoside, Methyl α-D-Mannopyranoside, ɑ-Methyl-D-mannopyranoside, (2R,3S,4S,5S,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

22925-5000 22925-0250 22925-1000
TS22925-5000EA 572 USD
TS22925-5000 TS22925-0250 TS22925-1000
Methyl-ɑ-D-mannopyranoside 99+%
Methyl-alpha-D-mannopyranoside
Formula: C₇H₁₄O₆
Melting Pt: 186…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00063262
CAS Number: 617-04-9

Order Now

Specification Test Results

Appearance White to off-white to light gray Powder
Infrared spectrum Conforms
Melting point 186 to 196 °C
Water ≤0.5 %
Sulfated ash ≤0.2 %
Solubility (10 % in water) Clear colorless
HPLC ≥99.0 % (ELSD)
Specific optical rotation +77° to +81° (20 °C, 589 nm) (c=10, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR